Heptanoyl chloride, 99%, Thermo Scientific Chemicals
Heptanoyl chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Heptanoyl chloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 164260250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2528-61-2
IUPAC Nameheptanoyl chloride
Molecular FormulaC7H13ClO
InChI KeyUCVODTZQZHMTPN-UHFFFAOYSA-N
SMILESCCCCCCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4290 to 1.4320 (20°C, 589 nm)
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only