Thermo Scientific Chemicals

Heptanoyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: 164265000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Heptanoyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: 164265000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 164265000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2528-61-2
IUPAC Name
heptanoyl chloride
Molecular Formula
C7H13ClO
InChI Key
UCVODTZQZHMTPN-UHFFFAOYSA-N
SMILES
CCCCCCC(Cl)=O
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Refractive index
1.4290 to 1.4320 (20°C, 589 nm)
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text