alpha-Pinene, 97%, stabilized with alpha-Tocopherol
alpha-Pinene, 97%, stabilized with alpha-Tocopherol
alpha-Pinene, 97%, stabilized with alpha-Tocopherol
alpha-Pinene, 97%, stabilized with alpha-Tocopherol
Thermo Scientific Chemicals

alpha-Pinene, 97%, stabilized with alpha-Tocopherol

CAS: 80-56-8 | C10H16 | 136.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 164360050
Price (USD)
26.65
Online Exclusive
29.90
Save 3.25 (11%)
Each
In stock
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
26.65
Online Exclusive
29.90
Save 3.25 (11%)
Each
Add to cart
Chemical Identifiers
CAS80-56-8
IUPAC Name2,6,6-trimethylbicyclo[3.1.1]hept-2-ene
Molecular FormulaC10H16
InChI KeyGRWFGVWFFZKLTI-UHFFFAOYNA-N
SMILESCC1=CCC2CC1C2(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4640 to 1.4680 (20°C, 589 nm)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0472366Certificate of AnalysisMay 17, 2025164362500, 164360010, 164360050
A0466561Certificate of AnalysisOct 02, 2024164362500, 164360010, 164360050
A0453000Certificate of AnalysisMay 07, 2024164362500, 164360010, 164360050
A0457829Certificate of AnalysisJan 31, 2024164362500, 164360010, 164360050
A0444845Certificate of AnalysisAug 25, 2023164362500, 164360010, 164360050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection