α-Pinene, 97%, stabilized with α-Tocopherol, Thermo Scientific Chemicals
α-Pinene, 97%, stabilized with α-Tocopherol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

α-Pinene, 97%, stabilized with α-Tocopherol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
1 kg
Packaging:
Glass bottle
Catalog number 164362500
Price (USD)/ Each
102.65
Online exclusive
114.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
102.65
Online exclusive
114.00 
Save 11.35 (10%)
Add to cart
α-Pinene, 97%, stabilized with α-Tocopherol, Thermo Scientific Chemicals
Catalog number164362500
Price (USD)/ Each
102.65
Online exclusive
114.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS80-56-8
IUPAC Name2,6,6-trimethylbicyclo[3.1.1]hept-2-ene
Molecular FormulaC10H16
InChI KeyGRWFGVWFFZKLTI-UHFFFAOYNA-N
SMILESCC1=CCC2CC1C2(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4640 to 1.4680 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only