Thermo Scientific Chemicals

1,3,5-Benzenetricarboxylic acid chloride, 98%, Thermo Scientific Chemicals

Catalog number: 164371000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

1,3,5-Benzenetricarboxylic acid chloride, 98%, Thermo Scientific Chemicals

Catalog number: 164371000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 164371000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4422-95-1
IUPAC Name
benzene-1,3,5-tricarbonyl trichloride
Molecular Formula
C9H3Cl3O3
InChI Key
UWCPYKQBIPYOLX-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC(=CC(=C1)C(Cl)=O)C(Cl)=O
Titration Argentometric
97.5 to 102.5 % (ex Cl)
Infrared spectrum
Conforms
Melting point
30°C to 35°C
Appearance (Form)
Solid or liquid
Appearance (Color)
White to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text