Thermo Scientific Chemicals

2-Bromobutyric acid, 99%, Thermo Scientific Chemicals

Catalog number: 164512500
250 mL, Glass Bottle, Each
Thermo Scientific Chemicals

2-Bromobutyric acid, 99%, Thermo Scientific Chemicals

Catalog number: 164512500
250 mL, Glass Bottle, Each
Quantity
Packaging
Catalog number: 164512500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
80-58-0
IUPAC Name
2-bromobutanoic acid
Molecular Formula
C4H7BrO2
InChI Key
YAQLSKVCTLCIIE-UHFFFAOYNA-N
SMILES
CCC(Br)C(O)=O
Appearance (Color)
Clear colorless to yellow
GC
>=98.5 %
Appearance (Form)
Liquid
Refractive index
1.4715 to 1.4755 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text