Thermo Scientific Chemicals

Betaine hydrochloride, 99%, extra pure, specified according to the requirements of U, Thermo Scientific Chemicals

Catalog number: 164550010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Betaine hydrochloride, 99%, extra pure, specified according to the requirements of U, Thermo Scientific Chemicals

Catalog number: 164550010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 164550010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
590-46-5
IUPAC Name
(carboxymethyl)trimethylazanium chloride
Molecular Formula
C5H12ClNO2
InChI Key
HOPSCVCBEOCPJZ-UHFFFAOYSA-N
SMILES
[Cl-].C[N+](C)(C)CC(O)=O
Infrared spectrum
Conforms (identification A)
Titration with HClO4
98.0 to 100.5 % (On anhydrous basis)
Residue after ignition
=<0.1 %
Appearance (Form)
Powder or crystals
Appearance (Color)
Colorless to white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text