Pyrogallol, 99%, Thermo Scientific Chemicals
Pyrogallol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pyrogallol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number 164560025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namebenzene-1,2,3-triol
Molecular FormulaC6H6O3
InChI KeyWQGWDDDVZFFDIG-UHFFFAOYSA-N
SMILESOC1=CC=CC(O)=C1O
Molecular Weight (g/mol)126.11
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to off-white
HPLC>=98.5 %
Appearance (Form)Crystalline powder
Melting point131°C to 135°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only