Thermo Scientific Chemicals

DL-Phenylalanine, 99%

Catalog number: 164575000
500 g, Plastic Bottle, Each
Thermo Scientific Chemicals

DL-Phenylalanine, 99%

Catalog number: 164575000
500 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 164575000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
150-30-1
IUPAC Name
2-amino-3-phenylpropanoic acid
Molecular Formula
C9H11NO2
InChI Key
COLNVLDHVKWLRT-UHFFFAOYNA-N
SMILES
NC(CC1=CC=CC=C1)C(O)=O
Appearance (Form)
Powder or crystals
Titration with HClO4
98.5 to 101.5 % (on dry substance)
Sulfated ash
=<0.3 %
Arsenic (As)
=<3 ppm
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text