L(-)-2-Octanol, 99+%, Thermo Scientific Chemicals
L(-)-2-Octanol, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L(-)-2-Octanol, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 164590100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name(2R)-octan-2-ol
Molecular FormulaC8H18O
InChI KeySJWFXCIHNDVPSH-MRVPVSSYSA-N
SMILESCCCCCC[C@@H](C)O
Molecular Weight (g/mol)130.23
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4250 to 1.4270 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=99.0 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only