Thermo Scientific Chemicals

DL-Camphor, 96%, Thermo Scientific Chemicals

Catalog number: 164640010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

DL-Camphor, 96%, Thermo Scientific Chemicals

Catalog number: 164640010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 164640010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
76-22-2
IUPAC Name
1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
Molecular Formula
C10H16O
InChI Key
DSSYKIVIOFKYAU-UHFFFAOYNA-N
SMILES
CC1(C)C2CCC1(C)C(=O)C2
Infrared spectrum
Conforms
Residue after evaporation
=<0.1 %
Appearance (Color)
White
Appearance (Form)
Crystals and/or chunks
Angle of optical rotation
-0.15° to +0.15° (20°C, 589 nm) (c=10, alcohol)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text