DL-Camphor, 96%, Thermo Scientific Chemicals
DL-Camphor, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DL-Camphor, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Packaging:
View all
Plastic drum
Catalog number 164640051
Price (USD)/ Each
197.65
Online exclusive
219.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
197.65
Online exclusive
219.00 
Save 21.35 (10%)
Add to cart
DL-Camphor, 96%, Thermo Scientific Chemicals
Catalog number164640051
Price (USD)/ Each
197.65
Online exclusive
219.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS76-22-2
IUPAC Name1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
Molecular FormulaC10H16O
InChI KeyDSSYKIVIOFKYAU-UHFFFAOYNA-N
SMILESCC1(C)C2CCC1(C)C(=O)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Residue after evaporation=<0.1 %
Appearance (Color)White
Appearance (Form)Crystals and/or chunks
Angle of optical rotation-0.15° to +0.15° (20°C, 589 nm) (c=10, alcohol)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only