2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%, Thermo Scientific Chemicals
2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 164650010
Price (USD)/ Each
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-ethyl-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC6H14O3
InChI KeyZJCCRDAZUWHFQH-UHFFFAOYSA-N
SMILESCCC(CO)(CO)CO
Molecular Weight (g/mol)134.18
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Flakes
Melting point>=57°C
Ash=<0.0015 %
Infrared spectrumConforms
Water=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only