4-Phenylpiperidine, 98%, Thermo Scientific Chemicals
4-Phenylpiperidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Phenylpiperidine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 164770010
Price (USD)/ Each
49.00
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
49.00
Add to cart
4-Phenylpiperidine, 98%, Thermo Scientific Chemicals
Catalog number164770010
Price (USD)/ Each
49.00
-
Add to cart
Chemical Identifiers
CAS771-99-3
IUPAC Name4-phenylpiperidine
Molecular FormulaC11H15N
InChI KeyUTBULQCHEUWJNV-UHFFFAOYSA-N
SMILESC1CC(CCN1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)Off-white to light brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
Melting point59°C to 64°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only