Thermo Scientific Chemicals

N,N'-Methylenebisacrylamide, ≥99%, for biochemistry, for electrophoresis

Catalog number: 164795000
500 g, Plastic Bottle, Each
Thermo Scientific Chemicals

N,N'-Methylenebisacrylamide, ≥99%, for biochemistry, for electrophoresis

Catalog number: 164795000
500 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 164795000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-26-9
IUPAC Name
N-[(prop-2-enamido)methyl]prop-2-enamide
Molecular Formula
C7H10N2O2
InChI Key
ZIUHHBKFKCYYJD-UHFFFAOYSA-N
SMILES
C=CC(=O)NCNC(=O)C=C
Appearance (Color)
White
Infrared spectrum
Conforms
UV
(1 % in water) 1 cm cell
Appearance (Form)
Crystalline powder or crystals or needles
Acrylic acid
=<0.05 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text