Triethoxysilane, 95%, Thermo Scientific Chemicals
Triethoxysilane, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethoxysilane, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 164812500
Price (USD)/ Each
454.65
Online exclusive
535.00 
Save 80.35 (15%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
454.65
Online exclusive
535.00 
Save 80.35 (15%)
Add to cart
Triethoxysilane, 95%, Thermo Scientific Chemicals
Catalog number164812500
Price (USD)/ Each
454.65
Online exclusive
535.00 
Save 80.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nametriethoxysilyl
Molecular FormulaC6H15O3Si
InChI KeyPKDCQJMRWCHQOH-UHFFFAOYSA-N
SMILESCCO[Si](OCC)OCC
Molecular Weight (g/mol)163.27
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.3770 to 1.3790 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
GC>=94 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only