Acrylamide, 98.5%, extra pure
Acrylamide, 98.5%, extra pure
Thermo Scientific Chemicals

Acrylamide, 98.5%, extra pure

Acrylamide, CAS # 79-06-1, is an organic amide compound used as a monomer in the synthesis of polyacrylamides.
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
10 kg
Packaging:
View all
Plastic Bottle
Catalog number 164830025
Price (USD)/ Each
234.65
Online exclusive
276.00 
Save 41.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
234.65
Online exclusive
276.00 
Save 41.35 (15%)
Add to cart
Acrylamide, 98.5%, extra pure
Catalog number164830025
Price (USD)/ Each
234.65
Online exclusive
276.00 
Save 41.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameprop-2-enamide
Molecular FormulaC3H5NO
InChI KeyHRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILESNC(=O)C=C
Molecular Weight (g/mol)71.08
View more
SpecificationsSpecification SheetSpecification Sheet
Color of solution(10 % in water) Clear and colorless
Appearance (Color)White
Melting point82°C to 86°C
Water=<2.0 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Acrylamide is an organic amide compound mainly used to as a monomer in the synthesis of polyacrylamide
  • Acrylamide is formed in foods containing carbohydrates in its composition, mainly when these foods are fried, grilled or baked

Applications

  • In laboratory experiments acrylamide participates in chemical reactions as an intermediate in the preparation of N-methylol acrylamide and N-butoxyacrylamide compounds
  • For molecular usage, this compound can be used as a carrier and is involved in the precipitation of small amounts of DNA
  • In molecular experiments it is used in gel electrophoresis, including western blot, and SDS-PAGE, to separate proteins and macromolecules based on their size and charge
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only