4-Chloro-2-phenylquinoline, 99%, Thermo Scientific Chemicals
4-Chloro-2-phenylquinoline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chloro-2-phenylquinoline, 99%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
2.5 g
Packaging:
Glass bottle
Catalog number 164870025
Price (USD)/ Each
-
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4979-79-7
IUPAC Name4-chloro-2-phenylquinoline
Molecular FormulaC15H10ClN
InChI KeyGLVDSTVYOFXBKT-UHFFFAOYSA-N
SMILESClC1=C2C=CC=CC2=NC(=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Melting point61°C to 64°C
Appearance (Color)Light yellow
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only