4-Hydroxy-3-methoxymandelic acid, 99%, Thermo Scientific Chemicals
4-Hydroxy-3-methoxymandelic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Hydroxy-3-methoxymandelic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 164900050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name(2S)-2-hydroxy-2-(4-hydroxy-3-methoxyphenyl)acetate
Molecular FormulaC9H9O5
InChI KeyCGQCWMIAEPEHNQ-QMMMGPOBSA-M
SMILESCOC1=CC(=CC=C1O)[C@H](O)C([O-])=O
Molecular Weight (g/mol)197.17
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=98.5 %
Appearance (Form)Crystalline powder
Melting point131°C to 134°C
Appearance (Color)White to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only