Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Use the Thermo Scientific™ MAbPac™ Capillary Reversed Phase HPLC Column high sensitivity HPLC or LC/MS characterization of intact proteins in top-down proteomics applications where sample amount is critically limited. This versatile column can be used for peptide-mapping characterization where the capillary format balances sensitivity with throughput. Additional applications are monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins. These columns have excellent resolution, long column lifetime, and low carry-over.
Column Format | Capillary HPLC Column |
Diameter (Metric) | 0.15 mm |
For Use With | High sensitivity HPLC or LC-MS of intact proteins in top-down proteomics with limited sample |
Length (Metric) | 150 mm |
Max. Pressure | 8700 psi (600 bar) |
Particle Shape | N/A |
Particle Size | 4 μm |
pH | 0 to 9 |
Pore Size | 1500 Å |
Surface Area | N/A |
Temperature | 110°C |
USP Type | N/A |
Stationary Phase | PSDVB (Polystyrene Divinylbenzene) |
Product Line | MAbPac |
Packing Material | PSDVB (Polystyrene Divinylbenzene) |
Column Type | Analytical HPLC Column |
Endcapped | N/A |
Phase | Reversed Phase |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Particle Size | Diameter (Metric) | Length (Metric) | Price (USD) | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
164947 | Each | 4 μm | 0.15 mm | 150 mm | 1,391.11 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Use the Thermo Scientific™ MAbPac™ Capillary Reversed Phase HPLC Column high sensitivity HPLC or LC/MS characterization of intact proteins in top-down proteomics applications where sample amount is critically limited. This versatile column can be used for peptide-mapping characterization where the capillary format balances sensitivity with throughput. Additional applications are monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins. These columns have excellent resolution, long column lifetime, and low carry-over.