2-Iodopropane, 98+%, stabilized, Thermo Scientific Chemicals
2-Iodopropane, 98+%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Iodopropane, 98+%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 164961000
Price (USD)/ Each
66.65
Online exclusive
78.80 
Save 12.15 (15%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
66.65
Online exclusive
78.80 
Save 12.15 (15%)
Add to cart
2-Iodopropane, 98+%, stabilized, Thermo Scientific Chemicals
Catalog number164961000
Price (USD)/ Each
66.65
Online exclusive
78.80 
Save 12.15 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-iodopropane
Molecular FormulaC3H7I
InChI KeyFMKOJHQHASLBPH-UHFFFAOYSA-N
SMILESCC(C)I
Molecular Weight (g/mol)169.99
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Refractive index1.4972 to 1.4992 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.0 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only