Ethyl caproate, 99+%, Thermo Scientific Chemicals
Ethyl caproate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl caproate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
500 mL
Packaging:
Glass bottle
Catalog number 164991000
Price (USD)/ Each
32.70
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
32.70
Add to cart
Ethyl caproate, 99+%, Thermo Scientific Chemicals
Catalog number164991000
Price (USD)/ Each
32.70
-
Add to cart
Chemical Identifiers
CAS123-66-0
IUPAC Nameethyl hexanoate
Molecular FormulaC8H16O2
InChI KeySHZIWNPUGXLXDT-UHFFFAOYSA-N
SMILESCCCCCC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4040 to 1.4090 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=99.0 %
Density(20°C) 0.867 to 0.871 kg/l
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only