UltraPure™ Agarose is a polysaccharide used for size-based separation of nucleic acids in agarose gel electrophoresis applications. UltraPure™ Agarose isRead more
Promotion
Promo code:P5780456
Gene Expression Promo
From RNA Isolation kits, reverse transcriptases to analysis of your gene with qPCR - upto 35% on all Gene Expression workflow productsLearn More
Have Questions?
Change view
Catalog Number
Quantity
16500100
100 g
16500500
500 g
2 Options
Catalog number 16500100
Price (USD)
239.65
Online Exclusive
261.00
Save 21.35 (8%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
239.65
Online Exclusive
261.00
Save 21.35 (8%)
Each
Add to cart
UltraPure™ Agarose is a polysaccharide used for size-based separation of nucleic acids in agarose gel electrophoresis applications. UltraPure™ Agarose is ideal for resolving DNA and RNA fragments from 100 bp to >30 kb. Features of UltraPure™ Agarose:
• Ideal for analysis and recovery of DNA and RNA for routine applications • Strong gel structure allows for better handling and less breakage • Can be used in protein electrophoresis applications such as Ouchterlony (antigen-antibody interaction assay) and radial immunodiffusion (RID) (antigen quantitation assay)
Improved packaging The new environmentally friendly packaging uses 75% less plastic than the original bottles. This means less energy and raw material used in manufacture, and less waste in landfills. Additionally, the easy-pour spout reduces the likelihood of spills and contamination.
Performance and quality testing The performance of UltraPure™ Agarose is evaluated to satisfy set specifications in appearance, moisture, gel strength, gelling temperature, melting temperature, sulfate content, electroendosmosis, and DNase/RNase activity.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Gel Electrophoresis
FormPowder
Gel CompatibilityAgarose Gels
Green FeaturesSustainable packaging
Melting PointStandard Melting Point
Product LineUltraPure™
Product TypeAgarose
Quantity100 g
Shipping ConditionRoom Temperature
Separation Range100 to >30000 bp
Unit SizeEach
Contents & Storage
Contents: 1 pouch containing 100 g of UltraPure™ Agarose Storage: 15°C to 30°C
Frequently asked questions (FAQs)
I want to pour my own gels. Which agarose should I use?
Our UltraPure Agarose is standard melting-point agarose designed for routine separation and analysis of DNA and RNA fragments in the 500-23,000 nt range. UltraPure Agarose 1000 is a specialized agarose that provides higher resolution of PCR fragments and other short DNA fragments. We also offer an UltraPure Low Melting Point Agarose, which is ideal for resolving DNA fragments from 10 to 1,000 bp with a low melting temperature of 65°C or less.