Ethyl caprate, 99+%, Thermo Scientific Chemicals
Ethyl caprate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl caprate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 165001000
Price (USD)/ Each
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS110-38-3
IUPAC Nameethyl decanoate
Molecular FormulaC12H24O2
InChI KeyRGXWDWUGBIJHDO-UHFFFAOYSA-N
SMILESCCCCCCCCCC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Density(20°C) 0.864 to 0.869 kg/l
Acid value=<1 mg KOH/g
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only