Methyl myristate, 99%, Thermo Scientific Chemicals
Methyl myristate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl myristate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
2.5 mL
500 mL
Packaging:
Glass bottle
Catalog number 165011000
Price (USD)/ Each
110.65
Online exclusive
123.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
110.65
Online exclusive
123.00 
Save 12.35 (10%)
Add to cart
Methyl myristate, 99%, Thermo Scientific Chemicals
Catalog number165011000
Price (USD)/ Each
110.65
Online exclusive
123.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Namemethyl tetradecanoate
Molecular FormulaC15H30O2
InChI KeyZAZKJZBWRNNLDS-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCC(=O)OC
Molecular Weight (g/mol)242.40
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4340 to 1.4380 (20°C, 589 nm)
Acid value=<1 mg KOH/g
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only