Methyl methoxyacetate, 99%, Thermo Scientific Chemicals
Methyl methoxyacetate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl methoxyacetate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 165091000
Price (USD)/ Each
79.50
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
79.50
Add to cart
Methyl methoxyacetate, 99%, Thermo Scientific Chemicals
Catalog number165091000
Price (USD)/ Each
79.50
-
Add to cart
Chemical Identifiers
CAS6290-49-9
IUPAC Namemethyl 2-methoxyacetate
Molecular FormulaC4H8O3
InChI KeyQRMHDGWGLNLHMN-UHFFFAOYSA-N
SMILESCOCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light brown
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
Refractive index1.3954 to 1.3974 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only