Thermo Scientific Chemicals

L(+)-Glutamic acid hydrochloride, 99%

Catalog number: 165230025
2.5 kg, Plastic Bottle, Each
Thermo Scientific Chemicals

L(+)-Glutamic acid hydrochloride, 99%

Catalog number: 165230025
2.5 kg, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 165230025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
138-15-8
IUPAC Name
hydrogen 2-aminopentanedioic acid chloride
Molecular Formula
C5H10ClNO4
InChI Key
RPAJSBKBKSSMLJ-UHFFFAOYNA-N
SMILES
[H+].[Cl-].NC(CCC(O)=O)C(O)=O
Loss on drying
=<0.5 % (105°C, 5 hrs)
Sulfated ash
=<0.1 %
Ammonium (NH4)
=<200 ppm
Iron (Fe)
=<10 ppm
pH
1 to 2.5 (2.5 % at 25°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text