Thermo Scientific Chemicals

N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 99+%, Thermo Scientific Chemicals

Catalog number: 165352500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 99+%, Thermo Scientific Chemicals

Catalog number: 165352500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 165352500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16357-59-8
Appearance (Color)
White to beige
Melting point
63°C to 66°C
Infrared spectrum
Conforms
HPLC
>=99.0 %
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text