2-Furoyl chloride, 98+%, Thermo Scientific Chemicals
2-Furoyl chloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Furoyl chloride, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 165380250
Price (USD)/ Each
59.50
-
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
59.50
Add to cart
2-Furoyl chloride, 98+%, Thermo Scientific Chemicals
Catalog number165380250
Price (USD)/ Each
59.50
-
Add to cart
Chemical Identifiers
CAS527-69-5
IUPAC Namefuran-2-carbonyl chloride
Molecular FormulaC5H3ClO2
InChI KeyOFTKFKYVSBNYEC-UHFFFAOYSA-N
SMILESClC(=O)C1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Titration RCOCl>=98.0 %
Refractive index1.5300 to 1.5320 (20°C, 589 nm)
Infrared spectrumConforms
Free acid (HCl)=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only