2-Furoyl chloride, 98+%
2-Furoyl chloride, 98+%
2-Furoyl chloride, 98+%
2-Furoyl chloride, 98+%
Thermo Scientific Chemicals

2-Furoyl chloride, 98+%

CAS: 527-69-5 | C5H3ClO2 | 130.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 165381000
Price (USD)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS527-69-5
IUPAC Namefuran-2-carbonyl chloride
Molecular FormulaC5H3ClO2
InChI KeyOFTKFKYVSBNYEC-UHFFFAOYSA-N
SMILESClC(=O)C1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Free acid (HCl)=<0.5 %
Impurity=<2.0 % Free carboxylic acid (as furoic acid)
GC>=98.0 %
Appearance (Form)Liquid
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0449251Certificate of AnalysisJan 31, 2025165381000, 165380250
A0464497Certificate of AnalysisJul 12, 2024165381000, 165380250
A0449251Certificate of AnalysisJan 12, 2024165381000, 165380250
A0431634Certificate of AnalysisJun 24, 2022165381000, 165380250
A0411099Certificate of AnalysisAug 09, 2021165381000, 165380250
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection