3-Diethylamino-1-propanol, 95%, Thermo Scientific Chemicals
3-Diethylamino-1-propanol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Diethylamino-1-propanol, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
Catalog number 165410500
Price (USD)/ Each
179.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
179.00
Add to cart
3-Diethylamino-1-propanol, 95%, Thermo Scientific Chemicals
Catalog number165410500
Price (USD)/ Each
179.00
-
Add to cart
Chemical Identifiers
CAS622-93-5
IUPAC Name3-(diethylamino)propan-1-ol
Molecular FormulaC7H17NO
InChI KeyWKCYFSZDBICRKL-UHFFFAOYSA-N
SMILESCCN(CC)CCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=94 %
Refractive index1.4425 to 1.4445 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only