Thermo Scientific Chemicals

Methyl disulfide, 99%, Thermo Scientific Chemicals

Catalog number: 165590025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Methyl disulfide, 99%, Thermo Scientific Chemicals

Catalog number: 165590025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 165590025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
624-92-0
IUPAC Name
(methyldisulfanyl)methane
Molecular Formula
C2H6S2
InChI Key
WQOXQRCZOLPYPM-UHFFFAOYSA-N
SMILES
CSSC
Appearance (Color)
Clear yellow
Infrared spectrum
Conforms
Refractive index
1.5243 to 1.5263 (20°C, 589 nm)
GC
>=98.5 %
Water
=<0.06 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text