Methyl disulfide, 99%, Thermo Scientific Chemicals
Methyl disulfide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl disulfide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 165590500
Price (USD)/ Each
34.80
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
34.80
Add to cart
Methyl disulfide, 99%, Thermo Scientific Chemicals
Catalog number165590500
Price (USD)/ Each
34.80
-
Add to cart
Chemical Identifiers
IUPAC Name(methyldisulfanyl)methane
Molecular FormulaC2H6S2
InChI KeyWQOXQRCZOLPYPM-UHFFFAOYSA-N
SMILESCSSC
Molecular Weight (g/mol)94.19
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Infrared spectrumConforms
Refractive index1.5243 to 1.5263 (20°C, 589 nm)
GC>=98.5 %
Water=<0.06 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only