Thermo Scientific Chemicals

5α-Cholestane, 98+%, Thermo Scientific Chemicals

Catalog number: 165602500
250 mg, Glass bottle, Each
Thermo Scientific Chemicals

5α-Cholestane, 98+%, Thermo Scientific Chemicals

Catalog number: 165602500
250 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 165602500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
481-21-0
IUPAC Name
9a,11a-dimethyl-1-(6-methylheptan-2-yl)-hexadecahydro-1H-cyclopenta[a]phenanthrene
Molecular Formula
C27H48
InChI Key
XIIAYQZJNBULGD-UHFFFAOYNA-N
SMILES
CC(C)CCCC(C)C1CCC2C3CCC4CCCCC4(C)C3CCC12C
Appearance (Form)
Crystalline powder
Melting point
78°C to 81°C
Specific optical rotation
+23° to +30° (20°C, 589 nm) (c=2, CHCl3)
Appearance (Color)
White
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text