Thermo Scientific Chemicals

6-Bromohexanoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 165675000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

6-Bromohexanoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 165675000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 165675000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4224-70-8
IUPAC Name
6-bromohexanoate
Molecular Formula
C6H10BrO2
InChI Key
NVRVNSHHLPQGCU-UHFFFAOYSA-M
SMILES
[O-]C(=O)CCCCCBr
Appearance (Color)
White to light orange
Appearance (Form)
Low melting crystals
Infrared spectrum
Conforms
Melting point
32°C to 35°C
Titration with NaOH
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text