Informational:Informational:This product ships via ground from US northeast.
Thermo Scientific Chemicals

DL-1,4-Dithiothreitol, 99%, for biochemistry

Catalog number: 165680250
25 g, Each
Thermo Scientific Chemicals

DL-1,4-Dithiothreitol, 99%, for biochemistry

Catalog number: 165680250
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 165680250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3483-12-3
IUPAC Name
(2R,3R)-1,4-disulfanylbutane-2,3-diol
Molecular Formula
C4H10O2S2
InChI Key
VHJLVAABSRFDPM-IMJSIDKUSA-N
SMILES
O[C@@H](CS)[C@@H](O)CS
Melting point
38°C to 43°C
Appearance (Color)
White
Appearance (Form)
Powder or beads
Infrared spectrum
Conforms
Titration Iodimetric
98.5 to 101.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • 1,4-Dithiothreitol (DTT or Cleland's reagent) reduces disulfide bonds in proteins, leading to loss of native conformation
  • DTT is not stable in solution and should be prepared in small aliquots prior to use; the use of EDTA in the solution can increase the half-life of DTT
  • Typically used at a final concentration of 0.1M for protein stabilization; recommended to create a 1M stock

Applications

  • Protects thiol groups from oxidation and acts as a chemical chelator in solution; maintains proteins in the denatured state
  • Common applications include SDS-page electrophoresis and protein isolation
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text