Isoamyl nitrite, 97%, stabilized, Thermo Scientific Chemicals
Isoamyl nitrite, 97%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isoamyl nitrite, 97%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
View all
Glass Bottle
Catalog number 165701000
Price (USD)/ Each
49.65
Online exclusive
55.30 
Save 5.65 (10%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
49.65
Online exclusive
55.30 
Save 5.65 (10%)
Add to cart
Isoamyl nitrite, 97%, stabilized, Thermo Scientific Chemicals
Catalog number165701000
Price (USD)/ Each
49.65
Online exclusive
55.30 
Save 5.65 (10%)
-
Add to cart
Chemical Identifiers
CAS110-46-3
IUPAC Name3-methylbutyl nitrite
Molecular FormulaC5H11NO2
InChI KeyOWFXIOWLTKNBAP-UHFFFAOYSA-N
SMILESCC(C)CCON=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.3850 to 1.3880 (20°C, 589 nm)
Stabilizer0.2 % Sodium carbonate
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only