Isoamyl nitrite, 97%, stabilized
Isoamyl nitrite, 97%, stabilized
Isoamyl nitrite, 97%, stabilized
Isoamyl nitrite, 97%, stabilized
Thermo Scientific Chemicals

Isoamyl nitrite, 97%, stabilized

CAS: 110-46-3 | C5H11NO2 | 117.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
2.5 L
500 mL
Packaging:
View all
Glass Bottle
Catalog number 165705000
Price (USD)
-
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS110-46-3
IUPAC Name3-methylbutyl nitrite
Molecular FormulaC5H11NO2
InChI KeyOWFXIOWLTKNBAP-UHFFFAOYSA-N
SMILESCC(C)CCON=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.3850 to 1.3880 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link