Decyl alcohol, 98+%
Decyl alcohol, 98+%
Decyl alcohol, 98+%
Decyl alcohol, 98+%
Thermo Scientific Chemicals

Decyl alcohol, 98+%

CAS: 112-30-1 | C10H22O | 158.28 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
檢視全部
Glass Bottle
產品號碼 165760010
價格 (TWD)
-
申請報價
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
化學識別
CAS112-30-1
IUPAC Namedecan-1-ol
Molecular FormulaC10H22O
InChI KeyMWKFXSUHUHTGQN-UHFFFAOYSA-N
SMILESCCCCCCCCCCO
檢視更多
規格Specification Sheet規格表
GC>=98.0 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4350 to 1.4390 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

文件與下載

證書

安全資料表

分享目錄編號、產品名稱或連結。

1x1 image pixel for data collection