9-Borabicyclo[3.3.1]nonane, 0.5M solution in THF, AcroSeal™
9-Borabicyclo[3.3.1]nonane, 0.5M solution in THF, AcroSeal™
9-Borabicyclo[3.3.1]nonane, 0.5M solution in THF, AcroSeal™
9-Borabicyclo[3.3.1]nonane, 0.5M solution in THF, AcroSeal™
Thermo Scientific Chemicals

9-Borabicyclo[3.3.1]nonane, 0.5M solution in THF, AcroSeal™

CAS: 280-64-8 | C8H15B | 122.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number 165818000
Price (USD)
365.00
Each
-
Add to cart
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
365.00
Each
Add to cart
Chemical Identifiers
CAS280-64-8
IUPAC Name9-borabicyclo[3.3.1]nonan-9-yl
Molecular FormulaC8H14B
InChI KeyAMKGKYQBASDDJB-UHFFFAOYSA-N
SMILES[B]1C2CCCC1CCC2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearancegently warm up and mix precipitate back into
Strength0.45 to 0.55 M (calc. molarity of 9-BBN based on active hydrogen)
Appearance (Color)Colorless to yellow
AppearanceMay develop some turbidity or precipitate
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid, Reduction reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link