Glycidyl methacrylate, 97%, stabilized
Glycidyl methacrylate, 97%, stabilized
Glycidyl methacrylate, 97%, stabilized
Glycidyl methacrylate, 97%, stabilized
Thermo Scientific Chemicals

Glycidyl methacrylate, 97%, stabilized

CAS: 106-91-2 | C7H10O3 | 142.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 165890025
Price (USD)
355.65
Online Exclusive
395.00
Save 39.35 (10%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
355.65
Online Exclusive
395.00
Save 39.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS106-91-2
IUPAC Name(oxiran-2-yl)methyl 2-methylprop-2-enoate
Molecular FormulaC7H10O3
InChI KeyVOZRXNHHFUQHIL-UHFFFAOYNA-N
SMILESCC(=C)C(=O)OCC1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.1 % (K.F.)
Refractive index1.4490 to 1.4510 (20°C, 589 nm)
Appearance (Form)Clear liquid
Color scale=<30 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection