Thermo Scientific Chemicals

Flufenamic acid, 97%

Catalog number: 165920500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Flufenamic acid, 97%

Catalog number: 165920500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 165920500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
530-78-9
IUPAC Name
2-{[3-(trifluoromethyl)phenyl]amino}benzoic acid
Molecular Formula
C14H10F3NO2
InChI Key
LPEPZBJOKDYZAD-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC=C1NC1=CC=CC(=C1)C(F)(F)F
Appearance (Form)
Crystalline powder and/or chunks
Infrared spectrum
Conforms
Appearance (Color)
White to light yellow to grey-green
Melting point
132°C to 137°C
Titration with NaOH
96.0 to 104.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text