Thermo Scientific Chemicals

4-Methyl-3-nitrobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 165990250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Methyl-3-nitrobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 165990250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 165990250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
96-98-0
IUPAC Name
4-methyl-3-nitrobenzoic acid
Molecular Formula
C8H7NO4
InChI Key
BBEWSMNRCUXQRF-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1[N+]([O-])=O)C(O)=O
Appearance (Color)
White to light yellow
Melting point
187°C to 190°C
Appearance (Form)
Crystalline powder
Titration with NaOH
>=98.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text