Thermo Scientific Chemicals

Thiazole, 99%, Thermo Scientific Chemicals

Catalog number: 166130050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Thiazole, 99%, Thermo Scientific Chemicals

Catalog number: 166130050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 166130050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
288-47-1
IUPAC Name
1,3-thiazole
Molecular Formula
C3H3NS
InChI Key
FZWLAAWBMGSTSO-UHFFFAOYSA-N
SMILES
S1C=CN=C1
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms
Refractive index
1.5365 to 1.5395 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text