Thermo Scientific Chemicals

L-Methionine, 98+%

Catalog number: 166161000
100 g, Plastic Bottle, Each
Thermo Scientific Chemicals

L-Methionine, 98+%

Catalog number: 166161000
100 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 166161000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63-68-3
IUPAC Name
(2S)-2-amino-4-(methylsulfanyl)butanoic acid
Molecular Formula
C5H11NO2S
InChI Key
FFEARJCKVFRZRR-BYPYZUCNSA-N
SMILES
CSCC[C@H](N)C(O)=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder or crystals
Loss on drying
=<0.5 % (1 g, 105°C)
Specific optical rotation
+22.4° to +24.7° (20°C, 589 nm) (c=2, 6 N HCl)
TLC
=<0.5 % of any individual impurity

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text