Acetonitrile-d3, for NMR, 99% atom D, Thermo Scientific Chemicals
Acetonitrile-d3, for NMR, 99% atom D, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetonitrile-d3, for NMR, 99% atom D, Thermo Scientific Chemicals

Acetonitrile-d3, C2H3N, CAS Number-2206-26-0, 2h3 acetonitrile, acetonitrile-2,2,2-d3, acetonitrile-d3, acetonitrile-d3, >-99.8 atom % d, acetonitrile-d3, 99.8 atom % d, acetonitrile-d isotopic 5g, cd3cn, methyl-d3 cyanide, trideuteroacetonitrile, 10mL, 02, IV, 428, 79.0 deg.C to 81.0 deg.C
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
10 mL
50 mL
250 mL
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 166230250
Price (USD)/ Each
141.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
166.00 
Save 24.35 (15%)
-
Add to cart
Quantity:
25 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
141.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
166.00 
Save 24.35 (15%)
Add to cart
Acetonitrile-d3, for NMR, 99% atom D, Thermo Scientific Chemicals
Catalog number166230250
Price (USD)/ Each
141.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
166.00 
Save 24.35 (15%)
-
Add to cart
Chemical Identifiers
CAS2206-26-0
IUPAC Name(²H₃)acetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-FIBGUPNXSA-N
SMILES[2H]C([2H])([2H])C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
D-Enrichment>=98.5 %
Appearance (Form)Liquid
Water=<0.1 % (Coulometric)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only