Chloroform-d, for NMR, 99.8% atom D, Thermo Scientific Chemicals
Chloroform-d, for NMR, 99.8% atom D, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chloroform-d, for NMR, 99.8% atom D, Thermo Scientific Chemicals

Chloroform-d, CHCl3, CAS Number-865-49-6, 2h chloroform, cdcl3, chloroform-d, chloroform, deutero, deuterated chloroform, deuterochloroform, methane-d, trichloro, trichloro deuterio methane, trichloromethane-d, unii-p1nw4885vt, 100mL, 01, IV, 54, 60.0 deg.C, CHEBI:85365, Colorless, 1.5000g/mL
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
25 mL
50 mL
100 mL
500 mL
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 166252500
Price (USD)/ Each
-
Quantity:
250 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nametrichloro(²H)methane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-MICDWDOJSA-N
SMILES[2H]C(Cl)(Cl)Cl
Molecular Weight (g/mol)120.38
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
D-Enrichment>=99.75 %
Water=<0.02 % (Coulometric)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only