Chloroform-d, for NMR, 99.8% atom D, contains 1 v/v% TMS, Thermo Scientific Chemicals
Chloroform-d, for NMR, 99.8% atom D, contains 1 v/v% TMS, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chloroform-d, for NMR, 99.8% atom D, contains 1 v/v% TMS, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
25 mL
100 mL
250 mL
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 166260500
Price (USD)/ Each
43.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
50.60 
Save 6.95 (14%)
-
Add to cart
Quantity:
50 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
43.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
50.60 
Save 6.95 (14%)
Add to cart
Chloroform-d, for NMR, 99.8% atom D, contains 1 v/v% TMS, Thermo Scientific Chemicals
Catalog number166260500
Price (USD)/ Each
43.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
50.60 
Save 6.95 (14%)
-
Add to cart
Chemical Identifiers
CAS865-49-6
IUPAC Nametrichloro(²H)methane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-MICDWDOJSA-N
SMILES[2H]C(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
D-Enrichment>=99.75 %
Appearance (Form)Liquid
TMS0.9 to 1.1 v/v%
Water=<0.02 % (Coulometric)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only