Thermo Scientific Chemicals

Chloroform-d, for NMR, 99.8% atom D, contains 1 v/v% TMS, Thermo Scientific Chemicals

Catalog number: 166261000
100 mL, Glass Bottle, Each
Thermo Scientific Chemicals

Chloroform-d, for NMR, 99.8% atom D, contains 1 v/v% TMS, Thermo Scientific Chemicals

Catalog number: 166261000
100 mL, Glass Bottle, Each
Quantity
Packaging
Catalog number: 166261000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
865-49-6
IUPAC Name
trichloro(²H)methane
Molecular Formula
CHCl3
InChI Key
HEDRZPFGACZZDS-MICDWDOJSA-N
SMILES
[2H]C(Cl)(Cl)Cl
Appearance (Color)
Clear colorless
D-Enrichment
>=99.75 %
Appearance (Form)
Liquid
TMS
0.9 to 1.1 v/v%
Water
=<0.02 % (Coulometric)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text