Chloroform-d, for NMR, 99.8 atom % D, contains 1 v/v% TMS
Chloroform-d, for NMR, 99.8 atom % D, contains 1 v/v% TMS
Chloroform-d, for NMR, 99.8 atom % D, contains 1 v/v% TMS
Chloroform-d, for NMR, 99.8 atom % D, contains 1 v/v% TMS
Thermo Scientific Chemicals

Chloroform-d, for NMR, 99.8 atom % D, contains 1 v/v% TMS

CAS: 865-49-6 | CHCl3 | 120.375 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
100 mL
25 mL
50 mL
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 166262500
Price (USD)
-
Quantity:
250 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS865-49-6
IUPAC Nametrichloro(²H)methane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-MICDWDOJSA-N
SMILES[2H]C(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
D-Enrichment>=99.75 %
Appearance (Form)Liquid
TMS0.9 to 1.1 v/v%
Water=<0.02 % (Coulometric)
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0472201Certificate of AnalysisMay 27, 2025166260500, 166262500, 166260250, 166261000
A0460247Certificate of AnalysisMay 17, 2025166260500, 166262500, 166260250, 166261000
A0451134Certificate of AnalysisMay 17, 2025166260500, 166262500, 166260250, 166261000
A0450948Certificate of AnalysisMay 17, 2025166260500, 166262500, 166260250, 166261000
A0469267Certificate of AnalysisMay 17, 2025166260500, 166262500, 166260250, 166261000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection